accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClCl

CAS RN 37680-73-2

IUPAC Name: 2,2',4,5,5'-pentachloro-1,1'-biphenyl
Synonyms: pentachlorobiphenyl
Molecular Weight (g/mol): 326.42
Molecular Formula: C12H5Cl5
InChi Key: LAHWLEDBADHJGA-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C(=C1)C1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results