Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2',4,5,5'-Pentachlorobiphenyl Solution (BZ No. 101), 100μg/mL in Hexane, Ultra Scientific

Catalog No. USRPC039S
Encompass
Change view
Click to view available options
Quantity:
1 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RPC-039S 1 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. US-RPC-039S Supplier Agilent Technologies Supplier No. RPC039S
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 37680-73-2 , 110-54-3
Molecular Formula C12H5Cl5
Molecular Weight (g/mol) 326.422
InChI Key LAHWLEDBADHJGA-UHFFFAOYSA-N
PubChem CID 37807
IUPAC Name 1,2,4-trichloro-5-(2,5-dichlorophenyl)benzene
SMILES C1=CC(=C(C=C1Cl)C2=CC(=C(C=C2Cl)Cl)Cl)Cl

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 x 2 mL
Concentration or Composition (by Analyte or Components) 100μg/mL (hexane)
Chemical Name or Material 2,2',4,5,5'-Pentachlorobiphenyl Solution (BZ No. 101), 100μg/mL (hexane)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.