accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3769-23-1

IUPAC Name: 4-methylhex-1-ene
Synonyms: 4-methyl-1-hexene
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: SUWJESCICIOQHO-UHFFFAOYNA-N
SMILES: CCC(C)CC=C
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)