Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1-hexene 99.0+%, TCI America™
SDP

Catalog No. M03025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03025ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M03025ML Supplier TCI America Supplier No. M03025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3769-23-1
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00039930
InChI Key SUWJESCICIOQHO-UHFFFAOYSA-N
PubChem CID 19589
IUPAC Name 4-methylhex-1-ene
SMILES CCC(C)CC=C

Specifications

Color Colorless
Boiling Point 86°C
Quantity 5 mL
UN Number 3295
Formula Weight 98.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-hexene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.