accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHNClHONOOOSOHOOSHOOO(E)

CAS RN 3769-62-8

IUPAC Name: (3E)-5-amino-3-[2-(3-chloro-2-hydroxy-5-nitrophenyl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonic acid
Synonyms: (3E)-5-amino-3-[2-(3-chloro-2-hydroxy-5-nitrophenyl)hydrazin-1-ylidene]-4-oxonaphthalene-2,7-disulfonic acid
Molecular Weight (g/mol): 518.85
Molecular Formula: C16H11ClN4O10S2
InChi Key: LODYRHRWPNPGLQ-ZHZULCJRSA-N
SMILES: NC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC(=CC(Cl)=C1O)[N+]([O-])=O)C(=C2)S(O)(=O)=O)S(O)(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results