Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallion, TCI America™
SDP

Catalog No. G02225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G02225G Supplier TCI America Supplier No. G02225G

Chemical Identifiers

CAS 3769-62-8
Molecular Formula C16H11ClN4O10S2
Molecular Weight (g/mol) 518.85
MDL Number MFCD00021475
InChI Key LODYRHRWPNPGLQ-ZHZULCJRSA-N
Synonym 8-Amino-2-(3-chloro-2-hydroxy-5-nitrophenylazo)-1-naphthol-3,6-disulfonic Acid
PubChem CID 44135574
IUPAC Name (3E)-5-amino-3-[2-(3-chloro-2-hydroxy-5-nitrophenyl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonic acid
SMILES NC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC(=CC(Cl)=C1O)[N+]([O-])=O)C(=C2)S(O)(=O)=O)S(O)(=O)=O

Specifications

Quantity 5 g
Formula Weight 518.85
Physical Form Crystalline Powder
Chemical Name or Material Gallion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.