accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFCl

CAS RN 37777-76-7

IUPAC Name: 2-(2-chloro-6-fluorophenyl)acetate
Synonyms: 2-(2-chloro-6-fluorophenyl)acetate
Molecular Weight (g/mol): 187.57
Molecular Formula: C8H5ClFO2
InChi Key: GUAIAAXDEJZRBP-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=C(F)C=CC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results