Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorophenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. C11265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11265G 5 g
C112625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11265G Supplier TCI America Supplier No. C11265G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37777-76-7
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.582
MDL Number MFCD00004319
InChI Key GUAIAAXDEJZRBP-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114
PubChem CID 123464
IUPAC Name 2-(2-chloro-6-fluorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)CC(=O)O)F

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 5 g
Formula Weight 188.58
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-fluorophenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.