accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37830-90-3

IUPAC Name: dimethyl-2H-1,3-dioxol-2-one
Synonyms: dimethyl-1,3-dioxol-2-one
Molecular Weight (g/mol): 114.1
Molecular Formula: C5H6O3
InChi Key: QYIOFABFKUOIBV-UHFFFAOYSA-N
SMILES: CC1=C(C)OC(=O)O1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results