Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethyl-1,3-dioxol-2-one 99.0+%, TCI America™
SDP

Catalog No. D192025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D192025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D192025G Supplier TCI America Supplier No. D192025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37830-90-3
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.10
MDL Number MFCD00135139
InChI Key QYIOFABFKUOIBV-UHFFFAOYSA-N
PubChem CID 142210
IUPAC Name dimethyl-2H-1,3-dioxol-2-one
SMILES CC1=C(C)OC(=O)O1

Specifications

Melting Point 78°C
Color Gray-White
Boiling Point 130°C
Quantity 25 g
Formula Weight 114.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dimethyl-1,3-dioxol-2-one
EINECSNumber (5)-6237
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.