accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3785-32-8

IUPAC Name: sodium 2-(2-carbamoylphenoxy)acetic acid
Synonyms: sodium salicylamide O-acetic acid
Molecular Weight (g/mol): 218.16
Molecular Formula: C9H9NNaO4
InChi Key: OEQSACUVTKYXIR-UHFFFAOYSA-N
SMILES: [Na+].NC(=O)C1=CC=CC=C1OCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results