Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium (2-Carbamoylphenoxy)acetate 98.0+%, TCI America™
SDP

Catalog No. C057925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C057925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C057925G Supplier TCI America Supplier No. C057925G

Chemical Identifiers

CAS 3785-32-8
Molecular Formula C9H8NNaO4
Molecular Weight (g/mol) 217.156
MDL Number MFCD00065100
InChI Key OEQSACUVTKYXIR-UHFFFAOYSA-M
Synonym Salicylamide O-Acetic Acid Sodium Salt, (2-Carbamoylphenoxy)acetic Acid Sodium Salt
PubChem CID 23682527
IUPAC Name sodium;2-(2-carbamoylphenoxy)acetate
SMILES C1=CC=C(C(=C1)C(=O)N)OCC(=O)[O-].[Na+]

Specifications

Melting Point 215°C
Color White
Quantity 25 g
Formula Weight 217.16
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium (2-Carbamoylphenoxy)acetate
EINECSNumber (9)-0486
RTECSNumber AF7952000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.