accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NNCl(E/Z)

CAS RN 3789-77-3

IUPAC Name: 4-[2-(3-chlorophenyl)diazen-1-yl]-N,N-dimethylaniline
Synonyms: 4-[2-(3-chlorophenyl)diazen-1-yl]-N,N-dimethylaniline
Molecular Weight (g/mol): 259.74
Molecular Formula: C14H14ClN3
InChi Key: ZWJXJMMRLCEJAL-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=CC(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results