Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Chloro-4-dimethylaminoazobenzene 97.0+%, TCI America™
SDP

Catalog No. C015825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C015825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C015825G Supplier TCI America Supplier No. C015825G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 3789-77-3
Molecular Formula C14H14ClN3
Molecular Weight (g/mol) 259.737
MDL Number MFCD00059480
InChI Key ZWJXJMMRLCEJAL-UHFFFAOYSA-N
Synonym 4-Dimethylamino-3′C-chloroazobenzene
PubChem CID 19625
IUPAC Name 4-[(3-chlorophenyl)diazenyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC(=CC=C2)Cl

Specifications

Melting Point 103°C
Quantity 25 g
Formula Weight 259.74
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Chloro-4-dimethylaminoazobenzene
RTECSNumber BX5040000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.