accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 379-57-7

IUPAC Name: tris(4-fluorophenyl)methanol
Synonyms: tris(4-fluorophenyl)methanol
Molecular Weight (g/mol): 314.31
Molecular Formula: C19H13F3O
InChi Key: JALVMPWRAMMHQY-UHFFFAOYSA-N
SMILES: OC(C1=CC=C(F)C=C1)(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results