Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4',4″-Trifluorotrityl Alcohol 98.0+%, TCI America™
SDP

Catalog No. T14661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14661G Supplier TCI America Supplier No. T14661G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 379-57-7
Molecular Formula C19H13F3O
Molecular Weight (g/mol) 314.31
MDL Number MFCD00191587
InChI Key JALVMPWRAMMHQY-UHFFFAOYSA-N
Synonym Tris(4-fluorophenyl)methanol
PubChem CID 2778005
IUPAC Name tris(4-fluorophenyl)methanol
SMILES OC(C1=CC=C(F)C=C1)(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1

Specifications

Melting Point 92°C
Color Yellow
Quantity 1 g
Formula Weight 314.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′,4′′-Trifluorotrityl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.