accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37939-50-7

IUPAC Name: heptane-1,2,7-triol
Synonyms: heptane-1,2,7-triol
Molecular Weight (g/mol): 148.2
Molecular Formula: C7H16O3
InChi Key: CTRCJSPDRXFNNN-UHFFFAOYNA-N
SMILES: OCCCCCC(O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results