Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,7-Heptanetriol 95.0+%, TCI America™
SDP

Catalog No. H09575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09575G Supplier TCI America Supplier No. H09575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37939-50-7
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.202
MDL Number MFCD01861288
InChI Key CTRCJSPDRXFNNN-UHFFFAOYSA-N
PubChem CID 19736292
IUPAC Name heptane-1,2,7-triol
SMILES C(CCC(CO)O)CCO

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 148.20
Percent Purity ≥95.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,2,7-Heptanetriol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.