accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37942-69-1

IUPAC Name: 4-butylcyclohexan-1-amine
Synonyms: 4-butylcyclohexan-1-amine
Molecular Weight (g/mol): 155.29
Molecular Formula: C10H21N
InChi Key: PZNZCCLRGXLYJI-UHFFFAOYSA-N
SMILES: CCCCC1CCC(N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results