Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylcyclohexylamine (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. B40145ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40145ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B40145ML Supplier TCI America Supplier No. B40145ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37942-69-1
Molecular Formula C10H21N
Molecular Weight (g/mol) 155.285
MDL Number MFCD09261273
InChI Key PZNZCCLRGXLYJI-UHFFFAOYSA-N
Synonym 1-Amino-4-butylcyclohexane
PubChem CID 425326
IUPAC Name 4-butylcyclohexan-1-amine
SMILES CCCCC1CCC(CC1)N

Specifications

Color Colorless
Quantity 5 mL
UN Number 2735
Formula Weight 155.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Butylcyclohexylamine (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.