accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFFFFFF

CAS RN 380-34-7

IUPAC Name: 1-ethoxy-1,1,2,3,3,3-hexafluoropropane
Synonyms: 1-ethoxy-1,1,2,3,3,3-hexafluoropropane
Molecular Weight (g/mol): 196.09
Molecular Formula: C5H6F6O
InChi Key: DMECHFLLAQSVAD-UHFFFAOYNA-N
SMILES: CCOC(F)(F)C(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results