accessibility menu, dialog, popup

UserName

Viewing profile as user:
SHHSSH

CAS RN 38004-59-0

IUPAC Name: benzene-1,3,5-trithiol
Synonyms: benzene-1,3,5-trithiol
Molecular Weight (g/mol): 174.29
Molecular Formula: C6H6S3
InChi Key: KXCKKUIJCYNZAE-UHFFFAOYSA-N
SMILES: SC1=CC(S)=CC(S)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results