accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONOOCH3H3CCH3OONOOH3CCH3H3COONOOH3CCH3H3COONOORhRh

CAS RN 380375-05-3

IUPAC Name: bis(λ²-rhodium(2+)) tetrakis(2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-3,3-dimethylbutanoate)
Synonyms: bis(λ²-rhodium(2+)) tetrakis(2-(1,3-dioxoisoindol-2-yl)-3,3-dimethylbutanoate)
Molecular Weight (g/mol): 1246.89
Molecular Formula: C56H56N4O16Rh2
InChi Key: PDTSLZCQKKXVQL-UHFFFAOYNA-J
SMILES: [Rh++].[Rh++].CC(C)(C)C(N1C(=O)C2=CC=CC=C2C1=O)C([O-])=O.CC(C)(C)C(N1C(=O)C2=CC=CC=C2C1=O)C([O-])=O.CC(C)(C)C(N1C(=O)C2=CC=CC=C2C1=O)C([O-])=O.CC(C)(C)C(N1C(=O)C2=CC=CC=C2C1=O)C([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results