Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis[N-phthaloyl-(R)-tert-leucinato]dirhodium Bis(ethyl Acetate) Adduct, TCI America™
SDP

Catalog No. T2054100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2054100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2054100MG Supplier TCI America Supplier No. T2054100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 380375-05-3
Molecular Formula C56H56N4O16Rh2
Molecular Weight (g/mol) 1246.89
MDL Number MFCD06797189
InChI Key PDTSLZCQKKXVQL-UHFFFAOYNA-J
Synonym tetrakis n-phthaloyl-r-tert-leucinato dirhodium bis ethyl acetate adduct, rhodium 2+ 2r-2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl-3,3-dimethylbutanoate-ethyl acetate 1/2/1
PubChem CID 131853186

Specifications

Quantity 100 mg
Formula Weight 1423.10
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis[N-phthaloyl-(R)-tert-leucinato]dirhodium Bis(ethyl Acetate) Adduct
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.