accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3H3COBOH3CH3CNH2

CAS RN 380430-65-9

IUPAC Name: hydrogen 1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine chloride
Synonyms: hydrogen 1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine chloride
Molecular Weight (g/mol): 269.58
Molecular Formula: C13H21BClNO2
InChi Key: BZZYJHQGJDCCCZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1(C)OB(OC1(C)C)C1=CC=CC(CN)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results