accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 380430-67-1

IUPAC Name: (2,3,6-trimethoxyphenyl)boronic acid
Synonyms: 2,3,6-trimethoxyphenylboronic acid
Molecular Weight (g/mol): 212.01
Molecular Formula: C9H13BO5
InChi Key: PBJYOPCURLYSAG-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(B(O)O)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results