accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3

CAS RN 380600-91-9

IUPAC Name: 9,9-dioctyl-9H-fluorene-2,7-dicarbaldehyde
Synonyms: 9,9-dioctylfluorene-2,7-dicarbaldehyde
Molecular Weight (g/mol): 446.68
Molecular Formula: C31H42O2
InChi Key: ZVQJNVBHJBLVSX-UHFFFAOYSA-N
SMILES: CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(C=O)=C2)C2=C1C=C(C=O)C=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results