Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Di-n-octylfluorene-2,7-dicarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. D46711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D46711G Supplier TCI America Supplier No. D46711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 380600-91-9
Molecular Formula C31H42O2
Molecular Weight (g/mol) 446.675
InChI Key ZVQJNVBHJBLVSX-UHFFFAOYSA-N
PubChem CID 53427856
IUPAC Name 9,9-dioctylfluorene-2,7-dicarbaldehyde
SMILES CCCCCCCCC1(C2=C(C=CC(=C2)C=O)C3=C1C=C(C=C3)C=O)CCCCCCCC

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 1 g
Formula Weight 446.68
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Di-n-octylfluorene-2,7-dicarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.