accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3(E/Z)

CAS RN 381716-24-1

IUPAC Name: 1-(diethoxymethyl)-4-(2-phenylethenyl)benzene
Synonyms: 1-(diethoxymethyl)-4-(2-phenylethenyl)benzene
Molecular Weight (g/mol): 282.38
Molecular Formula: C19H22O2
InChi Key: LKUHPNYFNZOTGE-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=CC=C(C=CC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results