Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Diethoxymethyl)-trans-stilbene 98.0+%, TCI America™
SDP

Catalog No. F04165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04165G Supplier TCI America Supplier No. F04165G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 381716-24-1
Molecular Formula C19H22O2
Molecular Weight (g/mol) 282.38
MDL Number MFCD02093470
InChI Key LKUHPNYFNZOTGE-UHFFFAOYSA-N
Synonym 4-Formyl-trans-stilbene Diethyl Acetal
PubChem CID 70700030
IUPAC Name 1-(diethoxymethyl)-4-(2-phenylethenyl)benzene
SMILES CCOC(OCC)C1=CC=C(C=CC2=CC=CC=C2)C=C1

Specifications

Melting Point 86°C
Color White
Quantity 5 g
Formula Weight 282.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Diethoxymethyl)-trans-stilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.