accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38183-12-9

IUPAC Name: 4'-phenyl-3H,3'H-spiro[2-benzofuran-1,2'-furan]-3,3'-dione
Synonyms: fluorescamine
Molecular Weight (g/mol): 278.26
Molecular Formula: C17H10O4
InChi Key: ZFKJVJIDPQDDFY-UHFFFAOYNA-N
SMILES: O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12
Quantity 
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
Color 
  • (4)
  • (4)
Grade 
  • (4)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (7)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)
  • (1)
  • (3)