accessibility menu, dialog, popup

UserName

Viewing profile as user:
SN

CAS RN 38210-35-4

IUPAC Name: 2-(1-benzothiophen-2-yl)pyridine
Synonyms: 2-(1-benzothiophen-2-yl)pyridine
Molecular Weight (g/mol): 211.28
Molecular Formula: C13H9NS
InChi Key: NRSBAUDUBWMTGL-UHFFFAOYSA-N
SMILES: S1C(=CC2=CC=CC=C12)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results