accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38212-33-8

IUPAC Name: 1-(4-chlorophenyl)piperazine
Synonyms: para-chlorophenylpiperazine
Molecular Weight (g/mol): 196.68
Molecular Formula: C10H13ClN2
InChi Key: UNEIHNMKASENIG-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)N1CCNCC1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)