Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)piperazine, 97%

Catalog No. AAJ6027814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6027814 25 g
AAJ6027803 1 g
AAJ6027806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6027814 Supplier Thermo Scientific Chemicals Supplier No. J6027814
Add to Cart

A 5-HT-1 serotonin receptor agonist | CAS: 38212-33-8 | C10H13ClN2 | 196.68 g/mol

1-(4-Chlorophenyl)piperazine is used as a 5-HT-1 serotonin receptor agonist. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Chlorophenyl)piperazine is used as a 5-HT-1 serotonin receptor agonist. It is also used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents. Store under inert gas. Sensitive to carbon dioxide
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38212-33-8
Molecular Formula C10H13ClN2
Molecular Weight (g/mol) 196.68
MDL Number MFCD00044823
InChI Key UNEIHNMKASENIG-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl piperazine, piperazine, 1-4-chlorophenyl, 1-4-chloro-phenyl-piperazine, para-chlorophenylpiperazine, pcpp, 1-4-chlorophenyl-piperazine, p-chlorophenylpiperazine, 4-chlorophenyl piperazine, 1-4-chlorphenyl piperazin
PubChem CID 97478
IUPAC Name 1-(4-chlorophenyl)piperazine
SMILES ClC1=CC=C(C=C1)N1CCNCC1

Specifications

Melting Point 76°C to 80°C
Color White
Boiling Point 113°C
Quantity 25 g
Solubility Information It is soluble in water.
Formula Weight 196.7
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Chlorophenyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.