accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3HO

CAS RN 382141-68-6

IUPAC Name: 1-(3-methoxyphenoxy)propan-2-ol
Synonyms: 1-(3-methoxyphenoxy)propan-2-ol
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: GUOGSANXPRRCMX-UHFFFAOYNA-N
SMILES: COC1=CC(OCC(C)O)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results