Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Methoxyphenoxy)-2-propanol 90.0+%, TCI America™
SDP

Catalog No. M10615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10615G 5 g
M106125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10615G Supplier TCI America Supplier No. M10615G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 382141-68-6
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00191540
InChI Key GUOGSANXPRRCMX-UHFFFAOYSA-N
Synonym 3-(2-Hydroxypropoxy)anisole
PubChem CID 22036449
IUPAC Name 1-(3-methoxyphenoxy)propan-2-ol
SMILES CC(COC1=CC=CC(=C1)OC)O

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 182.22
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(3-Methoxyphenoxy)-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.