accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2

CAS RN 38237-76-2

IUPAC Name: 1-(4-aminophenyl)hexan-1-one
Synonyms: 4-aminohexanoylphenone
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: YDQJFPLRVSPPGE-UHFFFAOYSA-N
SMILES: CCCCCC(=O)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results