CAS RN 38245-37-3
IUPAC Name:
disamarium(3+) tricarbonate
Synonyms:
disamarium(3+) tricarbonate
Molecular Weight (g/mol):
480.74
Molecular Formula:
C3O9Sm2
InChi Key:
QCZFMLDHLOYOQJ-UHFFFAOYSA-H
SMILES:
[Sm+3].[Sm+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
1
–
1
of
1
results
1
Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)
CAS: 38245-37-3 Molecular Formula: C3O9Sm2 Molecular Weight (g/mol): 480.74 MDL Number: MFCD00150444 InChI Key: QCZFMLDHLOYOQJ-UHFFFAOYSA-H IUPAC Name: disamarium(3+) tricarbonate SMILES: [Sm+3].[Sm+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O