accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 382631-42-7

IUPAC Name: nonane-1,2,9-triol
Synonyms: nonane-1,2,9-triol
Molecular Weight (g/mol): 176.26
Molecular Formula: C9H20O3
InChi Key: CUUVVDHSUIKLPH-UHFFFAOYNA-N
SMILES: OCCCCCCCC(O)CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results