Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,9-Nonanetriol 97.0+%, TCI America™
SDP

Catalog No. N06525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06525G Supplier TCI America Supplier No. N06525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 382631-42-7
Molecular Formula C9H20O3
Molecular Weight (g/mol) 176.256
MDL Number MFCD01861292
InChI Key CUUVVDHSUIKLPH-UHFFFAOYSA-N
PubChem CID 19013996
IUPAC Name nonane-1,2,9-triol
SMILES C(CCCC(CO)O)CCCO

Specifications

Melting Point 55°C
Color White
Quantity 5 g
Formula Weight 176.26
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,9-Nonanetriol
EINECSNumber (2)-2391
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.