accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FF

CAS RN 3828-49-7

IUPAC Name: 1,2-difluoro-3-methylbenzene
Synonyms: 1,2-difluoro-3-methylbenzene
Molecular Weight (g/mol): 128.12
Molecular Formula: C7H6F2
InChi Key: ZNEHIDGAPGVZSA-UHFFFAOYSA-N
SMILES: CC1=CC=CC(F)=C1F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)