Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluorotoluene 98.0+%, TCI America™
SDP

Catalog No. D34975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34975G 5 g
D34971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34975G Supplier TCI America Supplier No. D34975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3828-49-7
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.12
MDL Number MFCD01631506
InChI Key ZNEHIDGAPGVZSA-UHFFFAOYSA-N
Synonym 2,3-difluorotoluene, 1,2-difluoro-3-methyl-benzene, benzene, 1,2-difluoro-3-methyl, 2,3-difluoro methyl benzene, pubchem1610, 2,3-difluoro toluene, acmc-209iz4, 2.3-difluoro methyl benzene, ksc493c6t
PubChem CID 2774228
IUPAC Name 1,2-difluoro-3-methylbenzene
SMILES CC1=CC=CC(F)=C1F

Specifications

Color Yellow
Boiling Point 119°C
Quantity 5 g
UN Number 1993
Formula Weight 128.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Difluorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.