accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3829-86-5

IUPAC Name: hydrogen 1,10-phenanthroline chloride
Synonyms: hydrogen phen chloride
Molecular Weight (g/mol): 216.67
Molecular Formula: C12H9ClN2
InChi Key: QPXDKQBBJCTNOY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)