Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phenanthroline HCl, MP Biomedicals

Catalog No. ICN216553 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216553 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216553 Supplier MP Biomedicals, Inc Supplier No. 0521655301

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3829-86-5
Molecular Formula C12H9ClN2
Molecular Weight (g/mol) 216.668
InChI Key QPXDKQBBJCTNOY-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrochloride, 1,10-phenanthroline, monohydrochloride, phenanthroline hydrochloride, phen hydrochloride, 1,10-diazaphenanthrene hydrochloride, 1,10-phenanthroline, hydrochloride 1:1, 66-71-7 parent, o-phenanthrolinehydrochloride, c12h8n2.hcl, 1,10-phenanthrolinium chloride
PubChem CID 73350
IUPAC Name 1,10-phenanthroline;hydrochloride
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.Cl

Specifications

Quantity 1 g
Formula Weight 234.69
Chemical Name or Material o-Phenanthroline HCL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.