accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3834-42-2

IUPAC Name: (1,1,2,2,3,3,3-heptafluoropropyl)trimethylsilane
Synonyms: (1,1,2,2,3,3,3-heptafluoropropyl)trimethylsilane
Molecular Weight (g/mol): 242.21
Molecular Formula: C6H9F7Si
InChi Key: PKLASFRWKXWKII-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results