Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl(heptafluoropropyl)silane 98.0+%, TCI America™
SDP

Catalog No. T35935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35935G 5 g
T35931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35935G Supplier TCI America Supplier No. T35935G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3834-42-2
Molecular Formula C6H9F7Si
Molecular Weight (g/mol) 242.21
MDL Number MFCD00216712
InChI Key PKLASFRWKXWKII-UHFFFAOYSA-N
Synonym Trimethyl(perfluoropropyl)silane
PubChem CID 4158915
IUPAC Name (1,1,2,2,3,3,3-heptafluoropropyl)trimethylsilane
SMILES C[Si](C)(C)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 g
UN Number 1993
Formula Weight 242.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl(heptafluoropropyl)silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.