accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClO

CAS RN 3839-48-3

IUPAC Name: 2-chlorocyclohepta-2,4,6-trien-1-one
Synonyms: 2-chlorocyclohepta-2,4,6-trien-1-one
Molecular Weight (g/mol): 140.57
Molecular Formula: C7H5ClO
InChi Key: MTHNMAUDCHXFMM-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=CC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results