Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorotropone 98.0+%, TCI America™
SDP

Catalog No. C26741G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26741G Supplier TCI America Supplier No. C26741G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3839-48-3
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
MDL Number MFCD00182497
InChI Key MTHNMAUDCHXFMM-UHFFFAOYSA-N
Synonym 2-Chloro-2,4,6-cycloheptatrien-1-one
PubChem CID 19691
IUPAC Name 2-chlorocyclohepta-2,4,6-trien-1-one
SMILES ClC1=CC=CC=CC1=O

Specifications

Melting Point 68°C
Color White-Yellow
Boiling Point 97°C
Quantity 1 g
Formula Weight 140.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorotropone
RTECSNumber GU4025000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.