accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFClBr

CAS RN 384-16-7

IUPAC Name: 2-bromo-1-chloro-3-(trifluoromethyl)benzene
Synonyms: 2-bromo-1-chloro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 259.45
Molecular Formula: C7H3BrClF3
InChi Key: GMUWNTUONIQRFP-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(Cl)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results