CAS RN 3840-18-4
IUPAC Name:
2-(2-methylphenoxy)aniline
Synonyms:
aniline, 2-(o-tolyloxy)-
Molecular Weight (g/mol):
199.25
Molecular Formula:
C13H13NO
InChi Key:
JYJPXACGURQSCB-UHFFFAOYSA-N
SMILES:
CC1=CC=CC=C1OC1=CC=CC=C1N
1
–
1
of
1
results
1
2-Amino-2'-methyldiphenyl Ether 97.0+%, TCI America™
CAS: 3840-18-4 Molecular Formula: C13H13NO Molecular Weight (g/mol): 199.253 MDL Number: MFCD00025167 InChI Key: JYJPXACGURQSCB-UHFFFAOYSA-N Synonym: 2-Aminophenyl o-Tolyl Ether, 2-(o-Tolyloxy)aniline PubChem CID: 19692 IUPAC Name: 2-(2-methylphenoxy)aniline SMILES: CC1=CC=CC=C1OC2=CC=CC=C2N